N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98% (by HPLC) (max.6% H₂O)

Supplier: Ambeed
Warning

A171345-100G A171345-10G A171345-500G A171345-5G A171345-25G
77029-034EA 35 USD
77029-034 77029-036 77029-038 77029-040 77301-124
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98% (by HPLC) (max.6% H₂O)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate

Caution: for Research Use Only

Formula: C₁₈H₁₇NO₄
MDL Number: MFCD00062960
CAS Number: 79990-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR