1-Bromo-3-chloro-5-fluorobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-fluorobromobenzene
A167274-100G
A167274-10G
A167274-500G
A167274-5G
A167274-25G
77286-644EA
38
USD
77286-644
77286-646
77286-648
77286-650
77300-884
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene
Caution: for Research Use Only
Formula:
C₆H₃BrClF |
MDL Number:
MFCD00070747 CAS Number: 33863-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...