2-Furonitrile 98%
Supplier: Ambeed
|
Danger
|
A163728-25G
A163728-5G
A163728-1G
103805-760EA
7
USD
103805-760
103844-022
77300-672
77300-674
77320-132
2-Furonitrile 98%
2-Furonitrile
Caution: for Research Use Only
Formula:
C₅H₃NO Boiling Pt: 146…148 °C (760 mm Hg) Flash Pt: 35 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003223 CAS Number: 617-90-3 UN: 2924 ADR: 3,8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...