2-Iodophenol 98%

Supplier: Ambeed
Warning

A159770-10G A159770-500G A159770-5G A159770-25G A159770-100G
76973-492EA 7 USD
76973-492 76973-494 76973-496 77300-412 77567-328
2-Iodophenol 98%
2-Iodophenol

Caution: For research use only.

Formula: C₆H₅IO
MDL Number: MFCD00013963
CAS Number: 533-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR