2-Iodo-9,9-dimethylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-9,9-dimethyl-9H-fluorene

A159268-10G A159268-1G A159268-25G A159268-5G
103805-236EA 9 USD
103805-236 103843-454 76871-970 77300-394 77319-330 77319-332
2-Iodo-9,9-dimethylfluorene 98%
2-Iodo-9,9-dimethylfluorene

Caution: for Research Use Only

Formula: C₁₅H₁₃I
MW: 320.17 g/mol
Boiling Pt: 376.5 °C (760 mm Hg)
Melting Pt: 63…65 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12024272
CAS Number: 144981-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR