Luteolin 98%

Supplier: Ambeed
Warning

A144892-100G A144892-10G A144892-500G A144892-1G A144892-25G A144892-5G
103803-466EA 6 USD
103803-466 103841-544 77247-090 77247-092 77247-094 77299-420 77299-422 77299-424
Luteolin 98%
Luteolin

Caution: for Research Use Only

Formula: C₁₅H₁₀O₆
Melting Pt: 330 °C
Flash Pt: 239.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017309
CAS Number: 491-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR