N-Boc-ethylenediamine 97%
Supplier: Ambeed
|
Danger
|
A144595-10G
A144595-5G
A144595-25G
103841-500EA
5
USD
103841-500
76767-958
77299-400
77316-752
N-Boc-ethylenediamine 97%
N-Boc-ethylenediamine
Caution: for Research Use Only
Formula:
C₇H₁₆N₂O₂ Boiling Pt: 252.7 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191871 CAS Number: 57260-73-8 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...