2-Chloro-6-iodoaniline 98%

Supplier: Ambeed
Warning

A134005-10G A134005-250MG A134005-25G A134005-1G A134005-5G
103801-952EA 27 USD
103801-952 103832-892 77258-558 77258-560 77298-676 77314-660 77314-662
2-Chloro-6-iodoaniline 98%
2-Chloro-6-iodoaniline

Caution: for Research Use Only

Formula: C₆H₅ClIN
MW: 253.4681 g/mol
Boiling Pt: 273.1 °C (760 mm Hg)
Melting Pt: 66…67 °C
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD08443948
CAS Number: 84483-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR