3-Hydroxy-p-anisaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
A122974-100G
A122974-10G
A122974-25G
A122974-500G
A122974-5G
A122974-1000G
103831-150EA
5
USD
103831-150
76810-622
76810-624
76810-626
76810-628
77297-906
77882-991
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde
Caution: for Research Use Only
Formula:
C₈H₈O₃ Boiling Pt: 308.1 °C (760 mm Hg) Melting Pt: 119.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003369 CAS Number: 621-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...