BRAND® cell-culture™ Hand Controller Aspiration Unit
Supplier: Brandtech
25950
77296-260EA 114 USD
77296-260
BRAND® cell-culture™ Hand Controller Aspiration Unit
Pumps Vacuum Aspiration Systems
This instrument enables the removal of surplus liquids and can be utilized with pastuer pipets, pipette tips, or BRAND™ QuikSip™ bottle-top aspirator.
  • Includes suction tube and three distinct adapters designed to accommodate pasteur pipettes, capillaries or pipette tips (100 µl)
  • The adapter can be rotated for effortless fitting of pipettes/pipette tips
For effective liquid aspiration, simply connect the instrument to a collection bottle and a vacuum pump. Alternatively, enhance the setup with the QuikSip™ BT-aspirator mechanical bottle-top aspirator, offering a convenient alternative to the traditional vacuum pump. When used correctly, the instrument is virtually maintenance-free, providing reliable performance for lab tasks.
Always ensure to use an empty pipette and take precautions to prevent any liquid from entering the instrument. Hold the instrument horizontally during operation.
The cell-culture™ aspiration controller is not autoclavable. However, the included tube and adapters are autoclavable for sterilization purposes.
Delivery information: Includes suction tube and three adapters.
Caution: This instrument is intended solely for the aspiration of liquids. Vapors from liquids may come into contact with the adapter and suction tube. Therefore, it is crucial never to use the instrument for liquids that may corrode PVC (polyvinyl chloride), PP (polypropylene), or SI (silicone).