4-(Dimethylamino)benzonitrile 98%

Supplier: Ambeed
Warning

A814924-100G A814924-25G
77293-862EA 183 USD
77293-862 77293-864
4-(Dimethylamino)benzonitrile 98%
4-(Dimethylamino)benzonitrile

Caution: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD00001815
CAS Number: 1197-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR