3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%
Supplier: Ambeed
|
Warning
|
A682823-100G
A682823-1G
A682823-25G
A682823-5G
77293-154EA
871
USD
77293-154
77293-156
77293-158
77293-160
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
Caution: for Research Use Only
Formula:
C₂₀H₁₈N₂O₂S |
MDL Number:
MFCD00041869 CAS Number: 38215-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...