4,6-Dimethoxyindole 97%

Supplier: Ambeed
Warning

A632011-1G
77292-960EA 260.01 USD
77292-960
4,6-Dimethoxyindole 97%
4,6-Dimethoxyindole

Caution: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2
MDL Number: MFCD00128213
CAS Number: 23659-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR