3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 98%

Supplier: Ambeed
Warning

Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

A575945-100MG A575945-1G A575945-250MG
77292-628EA 94 USD
77292-628 77292-630 77292-632
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 98%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

Caution: for Research Use Only

Formula: C₂₅H₂₇IN₂S₂
MDL Number: MFCD00075033
CAS Number: 53213-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR