2-Chlorotrityl chloride 97%

Supplier: Ambeed
Danger

A418044-25G
77291-376EA 7 USD
77291-376
2-Chlorotrityl chloride 97%
2-Chlorotrityl chloride

Caution: for Research Use Only

Formula: C₁₉H₁₄Cl₂
MDL Number: MFCD00040399
CAS Number: 42074-68-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR