SB-366791 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide

A387775-100MG A387775-10MG A387775-1MG A387775-25MG A387775-50MG A387775-5MG
77291-034EA 102 USD
77291-034 77291-036 77291-038 77291-040 77291-042 77291-044
SB-366791 98%
SB-366791

Caution: for Research Use Only

Formula: C₁₆H₁₄ClNO₂
MDL Number: MFCD01033818
CAS Number: 472981-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR