SB-366791 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide
A387775-100MG
A387775-10MG
A387775-1MG
A387775-25MG
A387775-50MG
A387775-5MG
77291-034EA
102
USD
77291-034
77291-036
77291-038
77291-040
77291-042
77291-044
SB-366791 98%
SB-366791
Caution: for Research Use Only
Formula:
C₁₆H₁₄ClNO₂ |
MDL Number:
MFCD01033818 CAS Number: 472981-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...