4-Hydroxy-6-methoxycoumarin 95%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-6-methoxy-2H-chromen-2-one

A356528-100MG A356528-1G A356528-250MG
77290-702EA 23 USD
77290-702 77290-704 77290-706
4-Hydroxy-6-methoxycoumarin 95%
4-Hydroxy-6-methoxycoumarin

Caution: for Research Use Only

Formula: C₁₀H₈O₄
MDL Number: MFCD00463526
CAS Number: 13252-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR