4-Hydroxy-6-methoxycoumarin 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydroxy-6-methoxy-2H-chromen-2-one
A356528-100MG
A356528-1G
A356528-250MG
77290-702EA
23
USD
77290-702
77290-704
77290-706
4-Hydroxy-6-methoxycoumarin 95%
4-Hydroxy-6-methoxycoumarin
Caution: for Research Use Only
Formula:
C₁₀H₈O₄ |
MDL Number:
MFCD00463526 CAS Number: 13252-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...