4-Aminophenyl beta-D-Galactopyranoside 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
(2S,3R,4S,5R,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
A305879-100MG
A305879-1G
A305879-250MG
77290-076EA
44
USD
77290-076
77290-078
77290-080
4-Aminophenyl beta-D-Galactopyranoside 95%
4-Aminophenyl beta-D-Galactopyranoside
Caution: for Research Use Only
Formula:
C₁₂H₁₇NO₆ |
MDL Number:
MFCD00067362 CAS Number: 5094-33-7 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...