1,2-Difluoro-4-nitrobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Difluoronitrobenzene
A279035-100G
A279035-10G
A279035-25G
A279035-500G
A279035-5G
77289-608EA
20
USD
77289-608
77289-610
77289-612
77289-614
77289-616
1,2-Difluoro-4-nitrobenzene 98%
1,2-Difluoro-4-nitrobenzene
Caution: for Research Use Only
Formula:
C₆H₃F₂NO₂ MW: 159.09 g/mol Boiling Pt: 80 |
MDL Number:
MFCD00007198 CAS Number: 369-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...