1,2-Difluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoronitrobenzene

A279035-100G A279035-10G A279035-25G A279035-500G A279035-5G
77289-608EA 20 USD
77289-608 77289-610 77289-612 77289-614 77289-616
1,2-Difluoro-4-nitrobenzene 98%
1,2-Difluoro-4-nitrobenzene

Caution: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80
MDL Number: MFCD00007198
CAS Number: 369-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR