(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate
A270153-1G
A270153-100G
A270153-10G
A270153-500G
A270153-25G
A270153-5G
103812-986EA
8
USD
103812-986
103852-892
77092-656
77289-418
77289-420
77289-422
77289-424
77305-044
77305-046
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Caution: for Research Use Only
Formula:
C₂₀H₁₃O₄P Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010045 CAS Number: 39648-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...