4-Benzyloxybenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Benzyloxy)phenyl)methanol

A236599-100G A236599-25G A236599-5G A236599-10G
77288-606EA 87 USD
77288-606 77288-608 77615-950 77748-412
4-Benzyloxybenzyl alcohol 95%
4-Benzyloxybenzyl alcohol

Caution: for Research Use Only

Formula: C₁₄H₁₄O₂
MDL Number: MFCD00004654
CAS Number: 836-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR