2,3,4-Trifluorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid
A222388-10G
A222388-25G
A222388-1G
77288-244EA
23
USD
77288-244
77288-246
77748-360
2,3,4-Trifluorophenylboronic acid 98%
2,3,4-Trifluorophenylboronic acid
Caution: for Research Use Only
Formula:
C₆H₄BF₃O₂ |
MDL Number:
MFCD01863168 CAS Number: 226396-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...