Nicotinic acid (Niacin) 98%

Supplier: Ambeed
Warning

A211899-1000G A211899-100G A211899-10G A211899-25G A211899-500G
77287-980EA 37 USD
77287-980 77287-982 77287-984 77287-986 77287-988
Nicotinic acid (Niacin) 98%
Nicotinic acid (Niacin)

Caution: for Research Use Only

Formula: C₆H₅NO₂
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR