Nicotinic acid (Niacin) 98%
Supplier: Ambeed
|
Warning
|
A211899-1000G
A211899-100G
A211899-10G
A211899-25G
A211899-500G
77287-980EA
37
USD
77287-980
77287-982
77287-984
77287-986
77287-988
Nicotinic acid (Niacin) 98%
Nicotinic acid (Niacin)
Caution: for Research Use Only
Formula:
C₆H₅NO₂ |
MDL Number:
MFCD00006391 CAS Number: 59-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...