1-Bromo-3-chloro-5-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-fluorobromobenzene

A167274-100G A167274-10G A167274-500G A167274-5G A167274-25G
77286-644EA 38 USD
77286-644 77286-646 77286-648 77286-650 77300-884
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene

Caution: for Research Use Only

Formula: C₆H₃BrClF
MDL Number: MFCD00070747
CAS Number: 33863-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR