4-Bromo-2-fluoroaniline 97%

Supplier: Ambeed
Warning

A155079-100G A155079-500G A155079-25G
77286-250EA 41 USD
77286-250 77286-252 77300-104
4-Bromo-2-fluoroaniline 97%
4-Bromo-2-fluoroaniline

Caution: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD00010221
CAS Number: 367-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR