5-Bromo-2-furoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid

A148467-1000G A148467-100G A148467-10G A148467-1G A148467-25G A148467-500G A148467-5G
77286-028EA 297 USD
77286-028 77286-030 77286-032 77286-034 77286-036 77286-038 77286-040
5-Bromo-2-furoic acid 97%
5-Bromo-2-furoic acid

Caution: for Research Use Only

Formula: C₅H₃BrO₃
MW: 190.98 g/mol
MDL Number: MFCD00003239
CAS Number: 585-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR