5-Bromo-2-furoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid
A148467-1000G
A148467-100G
A148467-10G
A148467-1G
A148467-25G
A148467-500G
A148467-5G
77286-028EA
297
USD
77286-028
77286-030
77286-032
77286-034
77286-036
77286-038
77286-040
5-Bromo-2-furoic acid 97%
5-Bromo-2-furoic acid
Caution: for Research Use Only
Formula:
C₅H₃BrO₃ MW: 190.98 g/mol |
MDL Number:
MFCD00003239 CAS Number: 585-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...