6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
Supplier: Ambeed
|
Warning
|
A123399-10G
A123399-25G
A123399-5G
A123399-250MG
103800-400EA
9
USD
103800-400
103831-236
77284-970
77284-972
77312-520
77614-970
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione
Caution: for Research Use Only
Formula:
C₃H₂BrN₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00154757 CAS Number: 4956-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...