2-(Diphenylphosphino)benzoic acid 97%

Supplier: Ambeed
Warning

A108256-100G A108256-10G A108256-25G A108256-1G A108256-5G
103797-314EA 5 USD
103797-314 103823-094 77025-570 77025-572 77025-574 77283-700 77309-936
2-(Diphenylphosphino)benzoic acid 97%
2-(Diphenylphosphino)benzoic acid

Caution: for Research Use Only

Formula: C₁₉H₁₅O₂P
Boiling Pt: 451 °C (760 mm Hg)
Melting Pt: 174…181 °C
Flash Pt: 226.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00674024
CAS Number: 17261-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR