3-Fluoro-4-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A101466-25G
A101466-5G
A101466-1G
A101466-10G
A101466-100G
103821-672EA
22
USD
103821-672
76765-176
77282-750
77570-108
77623-824
77876-783
3-Fluoro-4-hydroxybenzaldehyde 97%
3-Fluoro-4-hydroxybenzaldehyde
Caution: for Research Use Only
Formula:
C₇H₅FO₂ Boiling Pt: 225.6 °C (760 mm Hg) Melting Pt: 121…124 °C Flash Pt: 90.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016626 CAS Number: 405-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...