Methyl-5-hydroxynicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate
A100312-25G
A100312-5G
A100312-1G
103795-360EA
10
USD
103795-360
103821-064
76717-486
77296-634
77282-470
Methyl-5-hydroxynicotinate 98%
Methyl 5-hydroxynicotinate
Caution: for Research Use Only
Formula:
C₇H₇NO₃ Boiling Pt: 374.2 °C (760 mm Hg) Melting Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191521 CAS Number: 30766-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...