BradyPrinter S3700 Multicolor and Cut Sign and Label Printers with Label Materials Kit

Supplier: Brady Worldwide
176868 176869 176870 176871 176872 176873 176874
77282-364KT 6746.14 USD
77282-364 77282-366 77282-368 77282-370 77282-372 77282-374 77282-376
BradyPrinter S3700 Multicolor and Cut Sign and Label Printers with Label Materials Kit
Printers Label Printers
Printer optimized for 1-, 2- or 4-color panel ribbons for multicolor made easy - no calibrating, no threading, no waste (single-color ribbon only in kit). The S3700 empowers any user to create any way they choose with time-saving onboard label templates, intuitive PC labeling software.

  • Cutter type: XY cutter and auto cutter
  • Multi-color printing capability
  • 600 labels per day (max.)

Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points. Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5-in.-per-second print speed. Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app.

Color kit - Get all the colors and capabilities you need in one convenient kit to create safety labels, lean visuals, lockout tags, work-in-process labels and more that truly pop.
Starter kit - Create professional, impactful safety tags by adding your specific hazard information to these print-on, write-on tags. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for just about any industrial surface.

Lean kit - Repositionable magnetic labels and printable ToughStripe® floor tape are perfect for bin labeling, area marking and other lean/5S applications. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for just about any industrial surface.
GHS kit - Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions. Durable pre-printed GHS labels contain ultra-aggressive adhesive to withstand exposure to moisture, chemicals and handling.

ARC kit - Durable, low-shrink all weather permanent adhesive vinyl labels with ANSI WARNING header will last and help keep you compliant. Raised panel labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
Pipe kit - Simply tap, print, go with preprogrammed onboard label templates plus 600 barcodes and symbols. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for labeling pipes indoors and out.

M611 kit – Desktop and portable printer in one kit. The M611 lets you create, share and print up to 2" width labels at 2" per second print speed anywhere, anytime.

Delivery information: Printers with color labels includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continuous white vinyl label (B30C-4000-595-WT);(1) 4" Continuous blue vinyl label (B30C-4000-595-BL);(1) 4" Continuous green vinyl label (B30C-4000-595-GN);(1) 4" Continuous red vinyl label (B30C-4000-595-RD);(1) 4" Continuous yellow vinyl label (B30C-4000-595-YL);(1) Black ribbon cartridge (B30-R10000);(1) White ribbon cartridge (B30-R10000-WT).
Printer starter kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continuous white vinyl label (B30C-4000-595-WT);(1) 2.25" Continuous white vinyl label (B30C-2250-595-WT);(1) 1.125" Continuous white vinyl label (B30C-1125-595-WT);(1) 4" Continuous yellow vinyl label(B30C-4000-595-YL);(1) 2.25" Continuous yellow vinyl label (B30C-2250-595-YL;(1) 4×6" White vinyl label (B30-25-595-BLNKWT);(1) OSHA DANGER tags (B30-255-851-WHT);(2) Black ribbon cartridges (B30-R10000);(1) White ribbon cartridge (B30-R10000-WT);(1) Red ribbon cartridge (B30-R10000-RD).
Lean/5S label kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB flash drive;(1) 4" White vinyl continuous label (B30C-4000-595-WT);(1) 2.25" Continuous white vinyl label (B30C-2250-595-WT);(1) 4" Continuous yellow label (B30C-4000-595-YL);(1) 2.25" Continuous yellow vinyl label (B30C-2250-595-YL);(1) 3.85×2.25" White vinyl label (B30-219-595-BLNKWT);(1) 2.5" Coninuous magnetic polyester label (B30C-2500-509-WT);(1) 4.25" Continuous magnetic polyester label (B30C-4250-509-WT);(1) 2.25" Continous yellow polyester label (B30C-2250-483YL-KT);(1) Black ribbon cartridge (B30-R10000);(1) White ribbon cartridge (B30-R10000-WT);(1) Red ribbon cartridge (B30-R10000-RD);(1) Blue ribbon cartridge (B30-R10000-BL);(1) 4-color ribbon cartridge (B30-R10000-KRYG-8).
GHS labeling kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) Pre-printed GHS labels (B30-262-7569-CLP4B);(1) Pre-printed GHS labels (B30-260-7569-CLP4B);(2) Black ribbon cartridges (B30-R10000).
Arc flash labeling kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB flash drive;(1) ANSI WARNING vinyl labels (B30-25-595-ANSIWA);(1) Raised panel polyester labels (B30EP-174-593-WT);(1) 2.25" Continuous orange vinyl label (B30C-2250-595-OR);(2) Black ribbon cartridges (B30-R10000).
Pipe marking kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continous yellow vinyl labels (B30C-4000-595-YL);(1) 2.25" Continous vinyl label (B30C-2250-595-YL);(1) 4" Continous green vinyl labels (B30C-4000-595-GN);(1) 2.25" Continous green vinyl labels (B30C-2250-595-GN);(1) 4" Continous red vinyl (B30C-4000-595-RD);(1) 2.25" Continous red vinyl labels (B30C-2250-595-RD);(1) 4" Continous blue vinyl labels (B30C-4000-595-BL);(1) 2.25" Continuous blue vinyl labels (B30C-2250-595-BL);(1) 4" Continous white vinyl labels (B30C-4000-595-WT);(1) Black ribbon cartridge (B30-R10000);(3) White ribbon cartridges (B30-R10000-WT).
Desktop and portable printer includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continuous white vinyl labels (B30C-4000-595-WT);(1) 4" Continous yellow vinyl labels (B30C-4000-595-YL);(1) 2.25" Continous white vinyl labels (B30C-2250-595-WT);(1) 2.25" Continous yellow vinyl labels (B30C-2250-595-YL);(2) Black ribbon cartridges (B30-R10000);(1) 1" Continous white vinyl labels (M6C-1000-595-WT);(1) 1" Continuous yellow vinyl labels (M6C-1000-595-YL);(1) 0.5" Continuous white vinyl labels (M6C-500-595-WT);(1) 0.5" Continuous yellow vinyl labels (M6C-500-595-YL);(2) Black ribbon cartirdges (M61-R10010); M611 Bluetooth label printer (M611).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR