4-Iodobenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: p-Iodobenzonitrile

A986300-1000G A986300-100G A986300-10G A986300-25G A986300-500G A986300-5G
77282-118EA 657.01 USD
77282-118 77282-120 77282-122 77282-124 77282-126 77282-128
4-Iodobenzonitrile 98%
4-Iodobenzonitrile

Caution: for Research Use Only

Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285
MDL Number: MFCD00051310
CAS Number: 3058-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR