2-Nitroethanol 97%

Supplier: Ambeed
Danger

A957769-100G A957769-10G A957769-1G A957769-25G A957769-5G
77282-056EA 167 USD
77282-056 77282-058 77282-060 77282-062 77282-064
2-Nitroethanol 97%
2-Nitroethanol

Caution: for Research Use Only

Formula: C₂H₅NO₃
MW: 91.07 g/mol
Boiling Pt: 194
Melting Pt: -80
MDL Number: MFCD00007405
CAS Number: 625-48-9
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR