3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A822485-25G A822485-5G A822485-10G A822485-1G
103841-140EA 13 USD
103841-140 76845-376 76845-378 77281-504 77308-400
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₂BrClF₃N
Boiling Pt: 210.5 °C (760 mm Hg)
Melting Pt: 28…32 °C
Flash Pt: 98.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878432
CAS Number: 71701-92-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR