3-Hydroxythiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Hydroxybenzene-1-thiol, 3-Hydroxybenzenethiol, 3-Hydroxythiophenol, 1-Hydroxy-3-mercaptobenzene, 3-Mercaptophenol, m-Hydroxythiophenol, 3-Hydroxyphenyl mercaptan

A679885-1G A679885-25G A679885-5G
77280-702EA 20 USD
77280-702 77569-292 77569-294
3-Hydroxythiophenol 98%
3-Hydroxythiophenol

Caution: For research use only.

Formula: C₆H₆OS
MW: 126.18
Melting Pt: 16.5
Density: 1.237
MDL Number: MFCD01317829
CAS Number: 40248-84-8
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR