3,4-Diacetyl-2,5-hexanedione 98%

Supplier: Ambeed
Warning

A567140-1G
77280-080EA 68 USD
77280-080
3,4-Diacetyl-2,5-hexanedione 98%
3,4-Diacetyl-2,5-hexanedione

Caution: for Research Use Only

Formula: C₁₀H₁₄O₄
MDL Number: MFCD00031531
CAS Number: 5027-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR