3-Chlorobenzenesulfonyl chloride 98% (by GC)
Supplier: Ambeed
|
Danger
|
A561876-10G
A561876-1G
A561876-25G
A561876-5G
77280-048EA
22
USD
77280-048
77280-050
77280-052
77280-054
3-Chlorobenzenesulfonyl chloride 98% (by GC)
3-Chlorobenzenesulphonyl chloride
Caution: for Research Use Only
Formula:
C₆H₄Cl₂O₂S |
MDL Number:
MFCD00051697 CAS Number: 2888-06-4 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...