3-Chlorobenzenesulfonyl chloride 98% (by GC)

Supplier: Ambeed
Danger

A561876-10G A561876-1G A561876-25G A561876-5G
77280-048EA 22 USD
77280-048 77280-050 77280-052 77280-054
3-Chlorobenzenesulfonyl chloride 98% (by GC)
3-Chlorobenzenesulphonyl chloride

Caution: for Research Use Only

Formula: C₆H₄Cl₂O₂S
MDL Number: MFCD00051697
CAS Number: 2888-06-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR