2-Methylindoline 98%

Supplier: Ambeed
Warning

A460836-100G A460836-25G A460836-500G A460836-5G
77279-262EA 32 USD
77279-262 77279-264 77279-266 77279-268
2-Methylindoline 98%
2-Methylindoline

Caution: for Research Use Only

Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 228
MDL Number: MFCD00005706
CAS Number: 6872-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR