2-Methylindoline 98%
Supplier: Ambeed
|
Warning
|
A460836-100G
A460836-25G
A460836-500G
A460836-5G
77279-262EA
32
USD
77279-262
77279-264
77279-266
77279-268
2-Methylindoline 98%
2-Methylindoline
Caution: for Research Use Only
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 228 |
MDL Number:
MFCD00005706 CAS Number: 6872-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...