Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
A424990-1G
A424990-25G
A424990-5G
A424990-100G
A424990-10G
A424990-500G
A424990-1000G
103827-454EA
7
USD
103827-454
103838-504
76842-152
76842-154
76842-156
77279-012
77279-014
77279-016
77616-842
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
Caution: for Research Use Only
Formula:
C₃₈H₃₂N₂O₅ Melting Pt: 201…204 °C Flash Pt: 472.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077049 CAS Number: 132388-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...