2-Oxazolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

A342282-25G A342282-100G A342282-10G A342282-500G
103836-836EA 6 USD
103836-836 76839-874 77277-994 77277-996 77277-998
2-Oxazolidinone 98%
2-Oxazolidinone

Caution: for Research Use Only

Formula: C₃H₅NO₂
MW: 87.08 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Flash Pt: 165.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00005268
CAS Number: 497-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR