3-Methylhexanoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methylhexanoicacid

A312091-1G
77277-554EA 1019.7 USD
77277-554
3-Methylhexanoic acid 97%
3-Methylhexanoic acid

Caution: for Research Use Only

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
MDL Number: MFCD00870847
CAS Number: 3780-58-3
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR