Diethyl aminomalonate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-aminomalonate hydrochloride

A306980-100G A306980-500G A306980-1000G A306980-25G A306980-5G
103810-044EA 53 USD
103810-044 103835-664 77277-436 77277-438 77305-940 77305-942 77305-944
Diethyl aminomalonate hydrochloride 98%
Diethyl aminomalonate hydrochloride

Caution: for Research Use Only

Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
Melting Pt: 165…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012510
CAS Number: 13433-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR