2-Amino-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A303477-25G A303477-5G A303477-100G A303477-10G A303477-500G
103835-544EA 20 USD
103835-544 76838-232 76838-234 77277-372 77277-374 77277-376
2-Amino-5-fluoropyridine 98%
2-Amino-5-fluoropyridine

Caution: for Research Use Only

Formula: C₅H₅FN₂
MW: 112.11
Boiling Pt: 195 °C (760 mm Hg)
Melting Pt: 93…97 °C
Flash Pt: 71.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01861120
CAS Number: 21717-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR